Assay Details
Target Gene Details
Entrez Gene ID: | 56356 |
Gene Name: | glycolipid transfer protein |
Gene Aliases: |
1110001F24Rik, C76925, C77564 |
Location: |
Chr.5:114669490-114690935 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gltp | NM_019821.2 | 5 | 898 | NP_062795.2 |
AK003218.1 | 1 | 365 | BAB22649.1 | |
AK157313.1 | 5 | 885 | BAE34045.1 | |
AK185281.1 | 1 | 189 | ||
BC016584.1 | 5 | 898 | AAH16584.1 | |
BC053729.1 | 5 | 841 | AAH53729.1 |
More Information
Set Membership: |
Intragenic Exonic |