Assay Details
Target Gene Details
Entrez Gene ID: | 22178 |
Gene Name: | tyrosinase-related protein 1 |
Gene Aliases: |
Oca3, TRP-1, TRP1, Tyrp, b, brown, isa |
Location: |
Chr.4:80834208-80851736 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tyrp1 | NM_001282014.1 | 2 | 349 | NP_001268943.1 |
NM_001282015.1 | 2 | 343 | NP_001268944.1 | |
NM_031202.3 | 2 | 364 | NP_112479.1 | |
AK148332.1 | 2 | 349 | BAE28487.1 | |
AK148370.1 | 2 | 255 | BAE28511.1 | |
AK148441.1 | 2 | 255 | BAE28554.1 | |
AK185619.1 | 1 | 61 | ||
AK219128.1 | 1 | 61 | ||
AK219226.1 | 1 | 61 | ||
BC076598.1 | 2 | 347 | AAH76598.1 | |
X03687.1 | 2 | 249 | CAA27323.1 |
More Information
Set Membership: |
Intragenic Exonic |