Assay Details
Target Gene Details
Entrez Gene ID: | 12580 |
Gene Name: | cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4) |
Gene Aliases: |
C77269, INK4c, p18, p18-INK4c, p18-INK6, p18INK4c |
Location: |
Chr.4:109660876-109666756 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdkn2c | NM_001301368.1 | 3 | 1535 | NP_001288297.1 |
NM_007671.3 | 2 | 603 | NP_031697.1 | |
AK003933.1 | 2 | 520 | BAB23080.1 | |
AK157209.1 | 3 | 1434 | BAE34001.1 | |
BC027026.1 | 2 | 603 | AAH27026.1 | |
BF657811.1 | 1 | 288 | ||
BG085472.2 | 3 | 453 | ||
U19596.1 | 3 | 455 | AAC52193.1 |