Assay Details
Target Gene Details
Entrez Gene ID: | 12580 |
Gene Name: | cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4) |
Gene Aliases: |
C77269, INK4c, p18, p18-INK4c, p18-INK6, p18INK4c |
Location: |
Chr.4:109660876-109666756 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdkn2c | NM_001301368.1 | NP_001288297.1 | ||
NM_007671.3 | 1 | 58 | NP_031697.1 | |
AK003933.1 | BAB23080.1 | |||
AK087461.1 | 1 | 1369 | ||
AK157209.1 | BAE34001.1 | |||
BC027026.1 | 1 | 58 | AAH27026.1 | |
BG085472.2 | ||||
U19596.1 | AAC52193.1 |