Assay Details
Target Gene Details
Entrez Gene ID: | 214444 |
Gene Name: | CDK5 regulatory subunit associated protein 2 |
Gene Aliases: |
2900018K03Rik, an, mKIAA1633 |
Location: |
Chr.4:70217690-70410461 on Build GRCm38 |
Assay Gene Location: | Within Exon 33 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdk5rap2 | NM_145990.3 | 33 | 4969 | NP_666102.2 |
AK129411.1 | 33 | 4969 | BAC98221.1 | |
BC059253.1 | 21 | 3560 | AAH59253.1 | |
BC086696.1 | 33 | 4871 | ||
BC110364.1 | 5 | 501 | AAI10365.1 |