Assay Details
Target Gene Details
Entrez Gene ID: | 231871 |
Gene Name: | diacylglycerol lipase, beta |
Gene Aliases: |
E330036I19Rik |
Location: |
Chr.5:143464487-143504442 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Daglb | NM_144915.3 | 15 | 3031 | NP_659164.2 |
AK086747.1 | 15 | 2943 | BAC39734.1 | |
BE992098.1 | 1 | 227 |
More Information
Set Membership: |
Intragenic Exonic |