Assay Details
Target Gene Details
Entrez Gene ID: | 231510 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 9 |
Gene Aliases: |
1-AGPAT, 1-AGPAT 9, 4933407I02Rik, 4933408F15, 9, A230097K15Rik, GPAT-3, mGPAT3 |
Location: |
Chr.5:100845710-100899102 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agpat9 | NM_172715.3 | 4 | 668 | NP_766303.1 |
AK030171.1 | 5 | 853 | BAC26820.1 | |
AK138410.1 | 3 | 828 | BAE23647.1 | |
AK160261.1 | 4 | 613 | BAE35719.1 | |
BC096769.1 | 4 | 601 | AAH96769.1 | |
BC138228.1 | 3 | 421 | ||
BC145669.1 | 3 | 421 |
More Information
Set Membership: |
Intragenic Exonic |