Assay Details
Target Gene Details
Entrez Gene ID: | 213990 |
Gene Name: | ArfGAP with GTPase domain, ankyrin repeat and PH domain 3 |
Gene Aliases: |
AGAP-3, AV028425, Centg3, Crag, Crag., MRIP-1, cnt-g3 |
Location: |
Chr.5:24452177-24502047 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agap3 | NM_139153.2 | 17 | 2427 | NP_631892.2 |
AF459091.1 | 17 | 2427 | AAL68640.1 | |
BC031173.1 | 8 | 999 | AAH31173.1 | |
BM943884.1 | 4 | 492 |