Assay Details
Target Gene Details
Entrez Gene ID: | 242891 |
Gene Name: | chaperonin containing TCP1, subunit 8 (theta)-like 1 |
Gene Aliases: |
Gm443 |
Location: |
Chr.5:25516067-25518027 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cct8l1 | NM_198621.2 | 1 | 985 | NP_941023.1 |
AK133375.1 | 1 | 985 | BAE21621.1 | |
BC050797.1 | 1 | 902 | AAH50797.1 |
More Information
Set Membership: |
Intragenic Exonic |