Assay Details
Target Gene Details
Entrez Gene ID: | 14825 |
Gene Name: | chemokine (C-X-C motif) ligand 1 |
Gene Aliases: |
Fsp, Gro1, KC, Mgsa, N51, Scyb1, gro |
Location: |
Chr.5:90891245-90893121 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcl1 | NM_008176.3 | 4 | 854 | NP_032202.1 |
AF352782.1 | 1 | 365 | ||
AK140312.1 | 1 | 1779 | ||
BC037997.1 | 4 | 821 | AAH37997.1 | |
BQ031102.1 | 1 | 111 | ||
J04596.1 | 4 | 789 | AAA40131.1 |
More Information
Set Membership: |
Intragenic Exonic |