Assay Details
Target Gene Details
Entrez Gene ID: | 100434 |
Gene Name: | solute carrier family 44, member 1 |
Gene Aliases: |
2210409B22Rik, 4833416H08Rik, AW547365, CHTL1, CTL1, Cdw92 |
Location: |
Chr.4:53440413-53622478 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc44a1 | NM_133891.3 | 16 | 2371 | NP_598652.3 |
AY249865.1 | 16 | 2127 | AAP81042.1 | |
BC010258.1 | 4 | 565 | AAH10258.1 | |
BC025941.1 | 7 | 933 | AAH25941.1 | |
BC113167.1 | 16 | 2127 | ||
BC113169.1 | 16 | 2127 |
More Information
Set Membership: |
Intragenic Exonic |