Assay Details
Target Gene Details
Entrez Gene ID: | 13483 |
Gene Name: | dipeptidylpeptidase 6 |
Gene Aliases: |
B930011P16Rik, D5Buc3, D5Buc4, D5Buc5, DPP VI, Dpp-6, Gm1377, In(5)6H-p, Peplb, Rw |
Location: |
Chr.5:26817357-27727500 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dpp6 | NM_001198886.1 | NP_001185815.1 | ||
NM_010075.2 | NP_034205.1 | |||
NM_207282.3 | NP_997165.2 | |||
AF092507.1 | AAC97366.1 | |||
AK047025.1 | 1 | 1376 | BAC32942.1 | |
AK051708.1 | ||||
AK082186.1 | ||||
AK158964.1 | BAE34745.1 | |||
BC085154.1 | AAH85154.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |