Assay Details
Target Gene Details
Entrez Gene ID: | 20404 |
Gene Name: | SH3-domain GRB2-like 2 |
Gene Aliases: |
9530001L19Rik, AI120490, AW555077, B930049H17Rik, EEN-B1, SH3PA, Sh3d2a |
Location: |
Chr.4:85205456-85389380 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sh3gl2 | NM_019535.2 | 9 | 1590 | NP_062408.2 |
AK044370.1 | 9 | 1605 | BAC31888.1 | |
AK134970.1 | 7 | 1301 | BAE22364.1 | |
AK139337.1 | 9 | 1555 | BAE23963.1 | |
BC018385.1 | 9 | 1590 | AAH18385.1 | |
U58886.1 | 9 | 1553 | AAC71774.1 |