Assay Details
Target Gene Details
Entrez Gene ID: | 18526 |
Gene Name: | protocadherin 10 |
Gene Aliases: |
6430521D13Rik, 6430703F07Rik, OL-pc, mKIAA1400 |
Location: |
Chr.3:45378398-45434579 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdh10 | NM_001098170.1 | NP_001091640.1 | ||
NM_001098171.1 | NP_001091641.1 | |||
NM_001098172.1 | NP_001091642.1 | |||
NM_011043.3 | 1 | 3864 | NP_035173.2 | |
AF334801.1 | AAK57195.1 | |||
AK034005.1 | BAC28544.1 | |||
AK036195.1 | ||||
AK038615.1 | ||||
AK039226.1 | BAC30284.1 | |||
AK046814.1 | ||||
AK081621.1 | ||||
AK081977.1 | ||||
AK122503.1 | BAC65785.1 | |||
AK158144.1 | 1 | 2371 | BAE34382.1 | |
BC065695.1 | 1 | 3651 | AAH65695.1 | |
U88549.1 | 1 | 3101 | AAD00651.1 |
More Information
Set Membership: |
Intragenic Exonic |