Assay Details
Target Gene Details
Entrez Gene ID: | 18526 |
Gene Name: | protocadherin 10 |
Gene Aliases: |
6430521D13Rik, 6430703F07Rik, OL-pc, mKIAA1400 |
Location: |
Chr.3:45378398-45434579 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdh10 | NM_001098170.1 | 4 | 3809 | NP_001091640.1 |
NM_001098171.1 | 4 | 3809 | NP_001091641.1 | |
NM_001098172.1 | 4 | 3809 | NP_001091642.1 | |
AA386527.1 | 2 | 433 | ||
AF334801.1 | 4 | 3045 | AAK57195.1 | |
AK018296.1 | BAB31151.1 | |||
AK034005.1 | 4 | 684 | BAC28544.1 | |
AK036195.1 | 4 | 2152 | ||
AK038615.1 | 4 | 2082 | ||
AK039226.1 | 4 | 1155 | BAC30284.1 | |
AK046814.1 | 4 | 711 | ||
AK081621.1 | 4 | 3808 | ||
AK081977.1 | 4 | 3805 | ||
AK122503.1 | 4 | 3778 | BAC65785.1 |
More Information
Set Membership: |
Intragenic Exonic |