Assay Details
Target Gene Details
Entrez Gene ID: | 12457 |
Gene Name: | CCR4 carbon catabolite repression 4-like (S. cerevisiae) |
Gene Aliases: |
AU043840, Ccr4, nocturnin |
Location: |
Chr.3:51224447-51251654 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccrn4l | NM_009834.2 | 3 | 2086 | NP_033964.1 |
AF183960.1 | 3 | 2072 | AAD56547.1 | |
AF199491.1 | 3 | 1579 | AAG01384.1 | |
AK041713.1 | 3 | 1602 | ||
AK084516.1 | 3 | 1607 | ||
AK087790.1 | 3 | 2086 | BAC40004.1 | |
AK147356.1 | 2 | 3940 | ||
AK147433.1 | 2 | 3940 | ||
AK149340.1 | 3 | 2064 | BAE28819.1 | |
BC139369.1 | 3 | 1725 | ||
BC139370.1 | 3 | 1725 | ||
U70139.1 | 3 | 8771 | AAB62717.1 |
More Information
Set Membership: |
Intragenic Exonic |