Assay Details
Target Gene Details
Entrez Gene ID: | 381572 |
Gene Name: | RIKEN cDNA 9430007A20 gene |
Gene Aliases: |
Aadacl4, Gm1034 |
Location: |
Chr.4:144519822-144529353 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
9430007A20Rik | NM_198662.3 | 4 | 1105 | NP_941064.2 |
AK034567.1 | 4 | 1106 | BAC28757.1 | |
BC148288.1 | 3 | 1010 |
More Information
Set Membership: |
Intragenic Exonic |