Assay Details
Target Gene Details
Entrez Gene ID: | 320024 |
Gene Name: | neutral cholesterol ester hydrolase 1 |
Gene Aliases: |
Aadacl1, B230106I24Rik, CPO-BP, Nceh |
Location: |
Chr.3:27183004-27245266 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nceh1 | NM_178772.3 | 5 | 3159 | NP_848887.1 |
AK034339.1 | 5 | 3086 | BAC28680.1 | |
AK045363.1 | 5 | 3160 | BAC32327.1 | |
AK128968.1 | 5 | 3170 | ||
AK135837.1 | 5 | 3149 | BAE22685.1 | |
AK159748.1 | 1 | 2102 | BAE35340.1 | |
AK159995.1 | 1 | 2102 | BAE35545.1 | |
BC082569.1 | 5 | 3142 | AAH82569.1 |
More Information
Set Membership: |
Intragenic Exonic |