Assay Details
Target Gene Details
Entrez Gene ID: | 269400 |
Gene Name: | regulator of telomere elongation helicase 1 |
Gene Aliases: |
AI451565, AW540478, Rtel |
Location: |
Chr.2:181288028-181356616 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rtel1 |
Target Gene Details
Entrez Gene ID: | 20262 |
Gene Name: | stathmin-like 3 |
Gene Aliases: |
9330161A03Rik, AI604839, R75323, Sclip |
Location: |
Chr.2:181306459-181314500 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stmn3 | NM_009133.3 | 5 | 1080 | NP_033159.1 |
AB007912.1 | 5 | 868 | BAA28629.1 | |
AF069708.1 | 5 | 849 | AAC21575.1 | |
AK002957.1 | 5 | 838 | BAB22479.1 | |
AK003203.1 | 5 | 838 | BAB22639.1 | |
AK003409.1 | 5 | 853 | BAB22773.1 | |
AK032737.1 | 5 | 1081 | BAC28003.1 | |
BC057017.1 | 5 | 856 | AAH57017.1 |