Assay Details
Target Gene Details
Entrez Gene ID: | 66665 |
Gene Name: | Myb/SANT-like DNA-binding domain containing 3 |
Gene Aliases: |
2410046L14Rik, 2810429P03Rik, 5730528L13Rik, Mysantd3 |
Location: |
Chr.4:48539832-48565892 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Msantd3 | NM_001145924.1 | 2 | 492 | NP_001139396.1 |
NM_001145925.1 | 2 | 353 | NP_001139397.1 | |
NM_028137.3 | 2 | 371 | NP_082413.2 | |
AK011381.1 | 2 | 494 | BAB27581.1 | |
AK013199.1 | 2 | 372 | BAB28708.1 | |
AK017795.1 | 2 | 496 | BAB30936.1 | |
AK075956.1 | 2 | 365 | BAC36081.1 | |
BC052696.1 | 2 | 414 | AAH52696.1 |