Assay Details
Target Gene Details
Entrez Gene ID: | 20760 |
Gene Name: | small proline-rich protein 2F |
Gene Aliases: |
- |
Location: |
Chr.3:92365187-92366442 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sprr2f | NM_011472.2 | 2 | 421 | NP_035602.1 |
AJ005564.1 | 1 | 382 | CAA06593.1 | |
AY158990.1 | 2 | 421 | AAN86827.1 |