Assay Details
Target Gene Details
Entrez Gene ID: | 108943 |
Gene Name: | tRNA methyltransferase 10A |
Gene Aliases: |
3110023L08Rik, AA794508, Rg9mtd2, Rnmtd2 |
Location: |
Chr.3:138143517-138159821 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trmt10a | NM_175389.4 | 8 | 3411 | NP_780598.2 |
AK146481.1 | 8 | 3543 | ||
BC090650.1 | 8 | 3411 | AAH90650.1 | |
BC141035.1 | 8 | 3399 |
More Information
Set Membership: |
Intragenic Exonic |