Assay Details
Target Gene Details
Entrez Gene ID: | 208188 |
Gene Name: | growth hormone secretagogue receptor |
Gene Aliases: |
C530020I22Rik |
Location: |
Chr.3:27371351-27378010 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ghsr | NM_177330.4 | 2 | 568 | NP_796304.1 |
AF332997.1 | 1 | 103 | AAG61141.1 | |
AK049671.1 | 2 | 569 | BAC33866.1 | |
AY056474.1 | 1 | 329 | AAL13336.1 | |
AY255570.1 | 1 | 246 | AAO85082.1 | |
BC120672.1 | 1 | 371 | ||
BC137882.1 | 1 | 371 |
More Information
Set Membership: |
Intragenic Exonic |