Assay Details
Target Gene Details
Entrez Gene ID: | 23831 |
Gene Name: | carbonic anhydrase 14 |
Gene Aliases: |
AW536446, Ca14 |
Location: |
Chr.3:95897768-95905118 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Car14 | NM_011797.2 | 1 | 277 | NP_035927.1 |
AB005450.1 | 1 | 277 | BAA78709.1 | |
AK009805.1 | 1 | 330 | BAC25271.1 | |
AK157764.1 | 1 | 277 | ||
BC046995.1 | 1 | 259 | AAH46995.1 |