Assay Details
Target Gene Details
Entrez Gene ID: | 71934 |
Gene Name: | carbonic anhydrase 13 |
Gene Aliases: |
2310075C21Rik, Ca13 |
Location: |
Chr.3:14641727-14663002 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Car13 | NM_024495.5 | 7 | 1016 | NP_078771.1 |
AF231123.1 | 7 | 966 | AAK16672.1 | |
AK010166.1 | 7 | 970 | BAB26742.1 | |
AK150871.1 | 7 | 968 | BAE29922.1 | |
AK150897.1 | 7 | 968 | BAE29942.1 | |
AK151519.1 | 7 | 954 | BAE30468.1 | |
AK151978.1 | 7 | 968 | BAE30845.1 | |
AK152124.1 | 7 | 965 | ||
AK153080.1 | 7 | 995 | BAE31705.1 | |
AK153258.1 | 7 | 956 | BAE31849.1 | |
AK153353.1 | 7 | 995 | BAE31927.1 | |
AK162621.1 | 7 | 1016 | BAE36996.1 | |
BC064050.1 | 7 | 967 | AAH64050.1 |