Assay Details
Target Gene Details
Entrez Gene ID: | 66077 |
Gene Name: | aurora kinase A interacting protein 1 |
Gene Aliases: |
0610033H09Rik, Aip, Akip, MRP-S38 |
Location: |
Chr.4:155831269-155833098 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aurkaip1 | NM_025338.4 | 3 | 775 | NP_079614.1 |
AK002737.1 | 3 | 394 | BAB22317.1 | |
AK003442.1 | 3 | 399 | BAB22792.1 | |
AK033452.1 | 3 | 382 | BAC28297.1 | |
AK144719.1 | 3 | 367 | BAE26030.1 | |
AK166049.1 | 3 | 379 | BAE38543.1 |