Assay Details
Target Gene Details
Entrez Gene ID: | 75173 |
Gene Name: | testis expressed 38 |
Gene Aliases: |
4930544O15Rik, Theg4 |
Location: |
Chr.4:115779831-115782759 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tex38 | NM_029196.1 | 2 | 182 | NP_083472.1 |
AK016041.1 | 2 | 167 | BAB30090.1 | |
CF982527.1 | 2 | 182 | ||
FJ468007.1 | 1 | 135 |