Assay Details
Target Gene Details
Entrez Gene ID: | 75540 |
Gene Name: | fucose-1-phosphate guanylyltransferase |
Gene Aliases: |
1700016E03Rik |
Location: |
Chr.3:155084915-155093378 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fpgt | NM_029330.2 | 4 | 873 | NP_083606.2 |
AJ276067.2 | 4 | 860 | CAC81971.2 | |
AK028640.1 | 4 | 870 | BAC26043.1 | |
AK045369.1 | 4 | 874 | ||
BC110551.1 | 4 | 873 | ||
BC110552.1 | 4 | 870 | AAI10553.1 |
More Information
Set Membership: |
Intragenic Exonic |