Assay Details
Target Gene Details
Entrez Gene ID: | 56046 |
Gene Name: | ubiquinol-cytochrome c reductase complex assembly factor 1 |
Gene Aliases: |
2310079L17Rik, 2410003P15Rik, 3110038N19Rik, Bfzb, Bfzp, Cbp3, Uqcc |
Location: |
Chr.2:155846886-155930343 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uqcc1 | NM_018888.4 | 10 | 2112 | NP_061376.2 |
NR_024487.2 | 10 | 2115 | ||
AF253516.1 | 10 | 1859 | AAF64519.1 | |
AK010376.1 | 10 | 1853 | BAB26894.1 | |
AK077323.1 | 10 | 1850 | BAC36749.1 | |
AK139488.1 | 10 | 2112 | BAE24034.1 | |
AK169413.1 | 10 | 1849 | BAE41159.1 | |
AK178882.1 | ||||
AK206360.1 | ||||
AK208091.1 | ||||
AK212618.1 | ||||
BC050884.1 | 10 | 2065 | AAH50884.1 | |
BC057570.1 | 10 | 2065 | AAH57570.1 |
More Information
Set Membership: |
Intragenic Exonic |