Assay Details
Target Gene Details
Entrez Gene ID: | 20672 |
Gene Name: | SRY (sex determining region Y)-box 18 |
Gene Aliases: |
AI385749, Ra, Ragl |
Location: |
Chr.2:181669837-181671640 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sox18 | NM_009236.2 | 2 | 1435 | NP_033262.2 |
AK144281.1 | 2 | 1436 | BAE25811.1 | |
AV167860.1 | 1 | 166 | ||
BC006612.1 | 2 | 1435 | AAH06612.1 | |
L35032.1 | 2 | 1404 | AAB18615.1 |
More Information
Set Membership: |
Intragenic Exonic |