Assay Details
Target Gene Details
Entrez Gene ID: | 27371 |
Gene Name: | SH2 domain containing 2A |
Gene Aliases: |
Lad, Ribp, Tsad |
Location: |
Chr.3:87846755-87855722 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sh2d2a | NM_001025571.2 | 1 | 173 | NP_001020742.1 |
NM_021309.3 | 1 | 173 | NP_067284.1 | |
AF203343.1 | 1 | 180 | AAF19396.1 |
More Information
Set Membership: |
Intragenic Exonic |