Assay Details
Target Gene Details
Entrez Gene ID: | 74838 |
Gene Name: | N(alpha)-acetyltransferase 15, NatA auxiliary subunit |
Gene Aliases: |
5730450D16Rik, 6330400I15, ASTBDN, Narg1, Tbdn-1, mNAT1 |
Location: |
Chr.3:51416016-51475985 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Naa15 | NM_053089.3 | 1 | 298 | NP_444319.3 |
AB221683.1 | ||||
AF510858.1 | 1 | 188 | AAO33713.1 | |
BC050017.1 | 1 | 189 | AAH50017.1 | |
BQ887090.1 | 1 | 293 | ||
CA327064.1 | 1 | 275 | ||
CX241349.1 | 1 | 297 |
More Information
Set Membership: |
Intragenic Exonic |