Assay Details
Target Gene Details
Entrez Gene ID: | 11641 |
Gene Name: | A kinase (PRKA) anchor protein 2 |
Gene Aliases: |
AA959716, AI649048, Akap-2, Akap-kl, B230340M18Rik, Prka2 |
Location: |
Chr.4:57845248-57896984 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Akap2 | NM_001035532.2 | 4 | 3400 | NP_001030609.1 |
NM_001035533.2 | 4 | 3380 | NP_001030610.1 | |
NM_009649.3 | 3 | 3221 | NP_033779.2 | |
AF033274.1 | 4 | 3782 | AAC02206.1 | |
AF033275.1 | 3 | 3743 | AAC02207.1 | |
AF033276.1 | 2 | 3564 | AAC02208.1 | |
AK147420.1 | 4 | 3368 | BAE27901.1 | |
AK162156.1 | 3 | 938 | ||
AK218394.1 | 1 | 102 |