Assay Details
Target Gene Details
Entrez Gene ID: | 329731 |
Gene Name: | family with sequence similarity 19, member A3 |
Gene Aliases: |
7530404M11Rik, TAFA3, Tafa-3 |
Location: |
Chr.3:104767405-104777547 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam19a3 | NM_183224.3 | 7 | 2648 | NP_899047.1 |
AK078681.1 | 7 | 2649 | ||
BC118531.1 | 7 | 2338 | ||
BE987068.1 | 1 | 110 | ||
BF461107.1 | 1 | 110 |
More Information
Set Membership: |
Intragenic Exonic |