Assay Details
Target Gene Details
Entrez Gene ID: | 21407 |
Gene Name: | transcription factor 15 |
Gene Aliases: |
Meso1, bHLH-EC2 |
Location: |
Chr.2:152143046-152149097 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tcf15 | NM_009328.2 | 2 | 661 | NP_033354.2 |
BC027533.1 | 2 | 661 | AAH27533.1 | |
U18658.1 | 2 | 701 | AAA86825.1 |
More Information
Set Membership: |
Intragenic Exonic |