Assay Details
Target Gene Details
Entrez Gene ID: | 15213 |
Gene Name: | hairy/enhancer-of-split related with YRPW motif 1 |
Gene Aliases: |
AI316788, AI414254, CHF2, HRT1, Herp2, Hesr1, bHLHb31, hesr-1 |
Location: |
Chr.3:8663359-8667038 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hey1 | NM_010423.2 | 5 | 1644 | NP_034553.2 |
AB041590.1 | 5 | 1537 | BAA95073.1 | |
AF151521.1 | 5 | 1592 | AAD38966.1 | |
AF172286.1 | 5 | 1625 | AAF14545.1 | |
AF176423.1 | 5 | 1606 | AAD51750.1 | |
AF232241.1 | 5 | 1624 | AAF37299.1 | |
AJ243895.1 | 5 | 1592 | CAB51321.1 | |
AK011149.1 | 5 | 1646 | BAB27432.1 | |
AK140212.1 | 5 | 1645 | BAE24283.1 | |
BC086635.1 | 5 | 1630 | AAH86635.1 |
More Information
Set Membership: |
Intragenic Exonic |