Assay Details
Target Gene Details
Entrez Gene ID: | 13636 |
Gene Name: | ephrin A1 |
Gene Aliases: |
AI325262, B61, Efl1, Epl1, Eplg1, Lerk1 |
Location: |
Chr.3:89271730-89280951 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Efna1 | NM_001162425.1 | 2 | 316 | NP_001155897.1 |
NM_010107.4 | 2 | 352 | NP_034237.3 | |
AK009144.1 | 2 | 349 | BAB26102.1 | |
AK079266.1 | 2 | 319 | ||
BC002046.1 | 2 | 317 | AAH02046.1 | |
CN696019.1 | 2 | 352 | ||
D38146.1 | 2 | 249 | BAA07344.1 | |
U26188.1 | 2 | 311 | AAA67563.1 | |
U90662.1 | 2 | 251 | AAB50237.1 |
More Information
Set Membership: |
Intragenic Exonic |