Assay Details
Target Gene Details
Entrez Gene ID: | 229003 |
Gene Name: | helicase with zinc finger 2, transcriptional coactivator |
Gene Aliases: |
PDIP1, Pric285, mKIAA1769, mPDIP1 |
Location: |
Chr.2:181227615-181242058 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Helz2 | NM_183162.2 | 7 | 3135 | NP_898985.2 |
AB201714.1 | 6 | 3039 | ||
AK157942.1 | 7 | 3134 | BAE34274.1 |
More Information
Set Membership: |
Intragenic Exonic |