Assay Details
Target Gene Details
Entrez Gene ID: | 70239 |
Gene Name: | general transcription factor IIIC, polypeptide 5 |
Gene Aliases: |
2700084A09Rik, TFIIIC63, TFiiiC2-63 |
Location: |
Chr.2:28566245-28583746 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtf3c5 | NM_001290484.1 | 12 | 2497 | NP_001277413.1 |
NM_148928.3 | 12 | 2479 | NP_683730.1 | |
AK012559.1 | 1 | 275 | ||
AK045419.1 | 12 | 2071 | BAC32354.1 | |
AK147014.1 | 12 | 2457 | BAE27608.1 | |
AK220093.1 | 1 | 133 | ||
AU016996.1 | 1 | 327 | ||
BC027055.1 | 12 | 2446 | AAH27055.1 | |
BC027247.1 | 12 | 2407 | AAH27247.1 | |
BC039171.1 | 12 | 2400 | AAH39171.1 |
More Information
Set Membership: |
Intragenic Exonic |