Assay Details
Target Gene Details
Entrez Gene ID: | 269252 |
Gene Name: | general transcription factor IIIC, polypeptide 4 |
Gene Aliases: |
5330400C03, AI426938, AU014771, AU017413, KAT12, TFIIIC90 |
Location: |
Chr.2:28822299-28840360 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtf3c4 | NM_001166033.1 | 1 | 283 | NP_001159505.1 |
NM_172977.3 | 1 | 283 | NP_766565.2 | |
AK030351.1 | 1 | 210 | BAC26916.1 | |
AK141269.1 | 1 | 264 | ||
AK146401.1 | 1 | 239 | BAE27142.1 | |
AK146403.1 | 1 | 239 | BAE27144.1 | |
AK150650.1 | 1 | 265 | BAE29737.1 | |
AK151179.1 | 1 | 265 | BAE30181.1 | |
AK153102.1 | 1 | 265 | BAE31722.1 | |
BC061476.1 | 1 | 216 | AAH61476.1 | |
BC095998.1 | 1 | 222 | AAH95998.1 | |
BC117919.1 | 1 | 195 | ||
BC117920.1 | 1 | 193 | ||
BU610856.1 | 1 | 210 | ||
CN530016.1 | 1 | 283 |
More Information
Set Membership: |
Intragenic Exonic |