Assay Details
Target Gene Details
Entrez Gene ID: | 72090 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 8 |
Gene Aliases: |
- |
Location: |
Chr.2:25080146-25085719 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Entpd8 | NM_028093.1 | 7 | 1126 | NP_082369.1 |
AK008590.1 | 3 | 648 | ||
AY364442.1 | 7 | 988 | AAQ84519.1 | |
BC031143.1 | 7 | 1122 |
More Information
Set Membership: |
Intragenic Exonic |