Assay Details
Target Gene Details
Entrez Gene ID: | 71532 |
Gene Name: | family with sequence similarity 217, member B |
Gene Aliases: |
9030418K01Rik, AI851938 |
Location: |
Chr.2:178414534-178422181 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam217b | NM_001081289.1 | 4 | 1962 | NP_001074758.1 |
AK155442.1 | 4 | 1963 |