Assay Details
Target Gene Details
Entrez Gene ID: | 207182 |
Gene Name: | gamma-glutamyltransferase 7 |
Gene Aliases: |
1110017C11Rik, 6330563L03Rik, Ggtl3 |
Location: |
Chr.2:155490379-155514860 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ggt7 | NM_144786.2 | 15 | 2243 | NP_659035.2 |
AF332053.1 | AAK56082.1 | |||
AF332054.1 | AAK56083.1 | |||
AK032051.1 | 15 | 2241 | BAC27672.1 | |
AK131175.1 | 12 | 4266 | BAD21425.1 | |
AK177521.1 | 1 | 53 | ||
AK198071.1 | ||||
AK203830.1 | 1 | 175 | ||
BC005772.1 | 15 | 2224 | AAH05772.1 |
More Information
Set Membership: |
Intragenic Exonic |