Assay Details
Target Gene Details
Entrez Gene ID: | 394436 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A1 |
Gene Aliases: |
Gnt1, UDPGT 1-1, UGT1A01, Udpgt-1a, UgtBr1 |
Location: |
Chr.1:88211959-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt1a1 | NM_201645.2 | 5 | 1982 | NP_964007.2 |
AY227194.1 | 5 | 1982 | AAP48593.1 | |
BC093516.1 | 5 | 1944 | AAH93516.1 | |
L02333.1 | 5 | 1954 | ||
S64760.1 | 5 | 1954 | AAB26033.2 |
Target Gene Details
Entrez Gene ID: | 394430 |
Gene Name: | UDP glycosyltransferase 1 family, polypeptide A10 |
Gene Aliases: |
A13, Ugt1a13 |
Location: |
Chr.1:88055411-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt1a10 | NM_201641.2 | 5 | 1995 | NP_964003.2 |
AY227201.1 | 5 | 1954 | AAP48600.1 | |
BC089569.1 | 5 | 1995 | AAH89569.1 |
Target Gene Details
Entrez Gene ID: | 22236 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A2 |
Gene Aliases: |
UDPGT 1-2, Ugt1 |
Location: |
Chr.1:88200611-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt1a2 | NM_013701.3 | 5 | 1959 | NP_038729.1 |
AK002629.1 | 5 | 1960 | BAB22243.1 | |
AY227195.1 | 5 | 1969 | AAP48594.1 |
Target Gene Details
Entrez Gene ID: | 394433 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A5 |
Gene Aliases: |
- |
Location: |
Chr.1:88166012-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt1a5 | NM_201643.2 | 5 | 1961 | NP_964005.2 |
AB094481.1 | 5 | 1940 | BAD11305.1 | |
AY227196.1 | 5 | 1961 | AAP48595.1 | |
BC139093.1 | 5 | 1941 | ||
BC139094.1 | 5 | 1941 |
Target Gene Details
Entrez Gene ID: | 394435 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A6B |
Gene Aliases: |
A9', Ugt1a9 |
Location: |
Chr.1:88103257-88218998 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt1a6b | NM_201410.3 | 6 | 2050 | NP_958812.3 |
AY227198.1 | 6 | 2006 | AAP48597.1 | |
BC026561.1 | 7 | 2030 | AAH26561.1 |
Target Gene Details
Entrez Gene ID: | 394432 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A7C |
Gene Aliases: |
A10', A13, Ugt1a10 |
Location: |
Chr.1:88095001-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt1a7c | NM_201642.4 | 5 | 2047 | NP_964004.1 |
AK128918.1 | 5 | 1973 | BAC87656.1 | |
AK144599.1 | 5 | 1939 | BAE25958.1 | |
AK153157.1 | 5 | 1937 | BAE31767.1 | |
AK154973.1 | 5 | 1970 | ||
AK159496.1 | 5 | 1985 | ||
AK162317.1 | 5 | 1971 | ||
AK165465.1 | 5 | 1971 | BAE38202.1 | |
AY227199.1 | 5 | 1951 | AAP48598.1 | |
BC141205.1 | 5 | 1956 | ||
BC141206.1 | 5 | 1956 |
Target Gene Details
Entrez Gene ID: | 394434 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A9 |
Gene Aliases: |
UDPGT, UDPGT 1-9, UGT1-09, UGT1.9, UGTP4, Ugt1a12, mUGTBr/P |
Location: |
Chr.1:88070779-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt1a9 | NM_201644.2 | 5 | 1969 | NP_964006.2 |
AY227200.1 | 5 | 1948 | AAP48599.1 | |
BC138699.1 | 5 | 1948 | ||
BC146021.1 | 5 | 1948 | ||
L27122.1 | 5 | 1894 | AAA40524.1 |
More Information
Set Membership: |
Intragenic Exonic |