Assay Details
Target Gene Details
Entrez Gene ID: | 228812 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class U |
Gene Aliases: |
5430426F17Rik, BC028278, Cdc91l1 |
Location: |
Chr.2:155278243-155357424 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pigu | NM_001004721.1 | 12 | 1561 | NP_001004721.1 |
AK143517.1 | 12 | 1561 | BAE25411.1 | |
AK171988.1 | 12 | 1540 | BAE42761.1 | |
AK181615.1 | ||||
AW538039.2 | 1 | 61 | ||
BC028278.1 | 4 | 692 | AAH28278.1 |