Assay Details
Target Gene Details
Entrez Gene ID: | 14571 |
Gene Name: | glycerol phosphate dehydrogenase 2, mitochondrial |
Gene Aliases: |
AA408484, AI448216, AU021455, AW494132, GPDH, Gdm1, Gpd-m, Gpdh-m, TISP38 |
Location: |
Chr.2:57237678-57370719 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpd2 | NM_001145820.1 | 17 | 3494 | NP_001139292.1 |
NM_010274.3 | 17 | 3369 | NP_034404.3 | |
AK032044.1 | 1 | 1090 | ||
AK034353.1 | 17 | 3278 | BAC28685.1 | |
AK079336.1 | 1 | 755 | ||
AK080240.1 | 1 | 85 | ||
AK149851.1 | 1 | 1110 | BAE29123.1 |
More Information
Set Membership: |
Intragenic Exonic |