Assay Details
Target Gene Details
Entrez Gene ID: | 227580 |
Gene Name: | C1q-like 3 |
Gene Aliases: |
1110065A22Rik, AI661623, C1ql, C1qtnf13, CTRP13, K100 |
Location: |
Chr.2:13001887-13010864 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1ql3 | NM_153155.2 | 1 | 504 | NP_694795.1 |
BC024634.1 | 1 | 504 | AAH24634.1 | |
EU399230.1 | 1 | 487 |