Assay Details
Target Gene Details
Entrez Gene ID: | 228866 |
Gene Name: | PDX1 C-terminal inhibiting factor 1 |
Gene Aliases: |
2310022K11Rik, C20orf67, F730014I05Rik |
Location: |
Chr.2:164879368-164891437 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcif1 | NM_146129.3 | 17 | 2467 | NP_666241.1 |
AK089358.1 | 17 | 2468 | BAC40853.1 | |
AK210761.1 | ||||
AK211497.1 | ||||
BC031431.1 | 17 | 2436 | AAH31431.1 |