Assay Details
Target Gene Details
Entrez Gene ID: | 227736 |
Gene Name: | RIKEN cDNA 1700019L03 gene |
Gene Aliases: |
- |
Location: |
Chr.2:32777413-32784405 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700019L03Rik | NM_025619.1 | 9 | 1583 | NP_079895.1 |
AK006127.1 | 9 | 1607 | BAB24421.1 | |
BC118617.1 | 9 | 1583 | ||
BC121825.1 | 9 | 1583 |
Target Gene Details
Entrez Gene ID: | 329384 |
Gene Name: | peptidyl-tRNA hydrolase 1 homolog (S. cerevisiae) |
Gene Aliases: |
2210013M04Rik, AW046744 |
Location: |
Chr.2:32775738-32777593 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptrh1 | NM_178595.3 | 5 | 825 | NP_848710.1 |
AK075833.1 | 5 | 826 | BAC35994.1 | |
BC098218.1 | 5 | 865 | AAH98218.1 |
More Information
Set Membership: |
Intragenic Exonic |