Assay Details
Target Gene Details
Entrez Gene ID: | 20652 |
Gene Name: | sterol O-acyltransferase 1 |
Gene Aliases: |
8430426K15Rik, ACAT-1, AW550831, Acact, ald, hid |
Location: |
Chr.1:156428108-156474328 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Soat1 | NM_009230.3 | 16 | 3289 | NP_033256.2 |
AK052299.1 | 13 | 2846 | BAC34925.1 | |
AK087210.1 | 1 | 102 | ||
AK142486.1 | 16 | 3206 | BAE25081.1 | |
DQ903181.1 | 16 | 3206 |
More Information
Set Membership: |
Intragenic Exonic |